4-Acetyl-2,3-difluorophenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

18085-1G 18085-25G 18085-5G
76909-166EA 62.6 USD
76909-166 76909-168 76909-170
4-Acetyl-2,3-difluorophenylboronic acid ≥97% research grade
4-Acetyl-2,3-difluorophenylboronic acid
Formula: C₈H₇BF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD13181626
CAS Number: 1451390-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR