3-Chloro-5-fluoro-4-methoxyphenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

18083-1G 18083-25G 18083-5G
76909-152EA 84.88 USD
76909-152 76909-154 76909-156
3-Chloro-5-fluoro-4-methoxyphenylboronic acid ≥97% research grade
3-Chloro-5-fluoro-4-methoxyphenylboronic acid
Formula: C₇H₇BClFO₃
MW: 204.391 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13181625
CAS Number: 1451392-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR