4-Bromo-1-ethoxy-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

17988-1G 17988-25G 17988-5G
76909-144EA 81.69 USD
76909-144 76909-146 76909-148
4-Bromo-1-ethoxy-2-methylbenzene ≥97%
4-Bromo-1-ethoxy-2-methylbenzene
Formula: C₉H₁₁BrO
MW: 215.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12547896
CAS Number: 79636-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR