3-Chloro-5-methoxypyridine ≥95%

Supplier: AOBChem USA

Warning

17837-1G 17837-25G 17837-500MG 17837-5G
76909-132EA 138.98 USD
76909-132 76909-134 76909-136 76909-138
3-Chloro-5-methoxypyridine ≥95%
3-Chloro-5-methoxypyridine
Formula: C₆H₆ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD09910244
CAS Number: 95881-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR