4-Chloro-N,N-diethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-N,N-diethylbenzamide

17784-1G 17784-500MG 17784-5G
76909-120EA 455.13 USD
76909-120 76909-122 76909-124
4-Chloro-N,N-diethylbenzamide ≥97%
4-Chloro-N,N-diethylbenzamide
Formula: C₁₁H₁₄ClNO
MW: 211.689 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00584080
CAS Number: 7461-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR