4-(Aminocarbonyl)-3,5-difluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

17770-1G 17770-25G 17770-500MG 17770-5G
76909-108EA 290.69 USD
76909-108 76909-110 76909-112 76909-114
4-(Aminocarbonyl)-3,5-difluorophenylboronic acid ≥97%
4-(Aminocarbonyl)-3,5-difluorophenylboronic acid
Formula: C₇H₆BF₂NO₃
MW: 200.935 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18447592
CAS Number: 924279-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR