4-(Benzyloxy)-3-fluorophenol ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-(Benzyloxy)-3-fluorobenzenol

17733-1G 17733-25G 17733-5G
76909-084EA 62.6 USD
76909-084 76909-086 76909-088
4-(Benzyloxy)-3-fluorophenol ≥97% research grade
4-(Benzyloxy)-3-fluorophenol
Formula: C₁₃H₁₁FO₂
MW: 218.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01814875
CAS Number: 81228-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR