N-(4-Bromo-2-fluorobenzoyl)morpholine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(4-Bromo-2-fluorobenzoyl)morpholine

17690-1G 17690-25G 17690-5G
76909-058EA 88.06 USD
76909-058 76909-060 76909-062
N-(4-Bromo-2-fluorobenzoyl)morpholine ≥97%
N-(4-Bromo-2-fluorobenzoyl)morpholine

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrFNO₂
MW: 288.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09945068
CAS Number: 924642-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR