2-Fluoro-4-(methylcarbamoyl)phenylboronic acid 97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-4-(methylcarbamoyl)benzeneboronic acid, 4-(N-Methylaminocarbonyl)-2-fluorophenylboronic acid

17684-1G 17684-25G 17684-5G 17684-500MG
76909-044EA 117.76 USD
76909-044 76909-046 76909-048 77648-996
2-Fluoro-4-(methylcarbamoyl)phenylboronic acid 97%
2-Fluoro-4-(methylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD11856043
CAS Number: 874289-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR