(4-(Methoxymethoxy)-3,5-dimethylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-(Methoxymethoxy)-3,5-dimethylphenylboronic acid

17648-1G 17648-25G 17648-5G
76909-024EA 79.57 USD
76909-024 76909-026 76909-028
(4-(Methoxymethoxy)-3,5-dimethylphenyl)boronic acid ≥97%
(4-(Methoxymethoxy)-3,5-dimethylphenyl)boronic acid
Formula: C₁₀H₁₅BO₄
MW: 210.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11856041
CAS Number: 223128-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR