3-Bromo-5-ethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

17643-1G 17643-25G 17643-500MG 17643-5G
76909-012EA 150.65 USD
76909-012 76909-014 76909-016 76909-018
3-Bromo-5-ethoxybenzaldehyde ≥97%
3-Bromo-5-ethoxybenzaldehyde
Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20526371
CAS Number: 1451391-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR