4-[(1,1-Dimethylethoxy)carbonyl](phenylmethyl)aminophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

17453-1G 17453-25G 17453-500MG 17453-5G
76908-956EA 142.17 USD
76908-956 76908-958 76908-960 76908-962
4-[(1,1-Dimethylethoxy)carbonyl](phenylmethyl)aminophenylboronic acid ≥97%
4-[(1,1-Dimethylethoxy)carbonyl](phenylmethyl)aminophenylboronic acid
Formula: C₁₈H₂₂BNO₄
MW: 327.183 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17011979
CAS Number: 693792-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR