3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4,4,5,5-Tetramethyl-2-[3-(4-Methyl-1-piperazinylmethyl)phenyl]-1,3,2-dioxaborolane, 1-Methyl-4-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)piperazine, 3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid Pinacol ester, 1-Methyl-4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperazine

17395-25G 17395-5G
76908-950EA 392.54 USD
76908-950 76908-952
3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester ≥97%
3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester
Formula: C₁₈H₂₉BN₂O₂
MW: 316.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09266181
CAS Number: 883738-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR