3-Benzylcarbamoyl-5-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(Benzylcarbamoyl)-5-fluorophenylboronic acid

17335-1G 17335-25G 17335-5G
76908-938EA 88.06 USD
76908-938 76908-940 76908-942
3-Benzylcarbamoyl-5-fluorophenylboronic acid ≥97%
3-Benzylcarbamoyl-5-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₃BFNO₃
Storage Temperature: Ambient
MDL Number: MFCD08235058
CAS Number: 874219-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR