3-Carbamoyl-5-fluorobenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Carbamoyl-5-fluorophenyl)boronic acid, 3-(Aminocarbonyl)-5-fluorophenylboronic acid

17329-1G 17329-25G 17329-500MG 17329-5G
76908-912EA 160.2 USD
76908-912 76908-914 76908-916 76908-918
3-Carbamoyl-5-fluorobenzeneboronic acid ≥97%
3-Carbamoyl-5-fluorobenzeneboronic acid
Formula: C₇H₇BFNO₃
MW: 182.95 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08056357
CAS Number: 871332-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR