3-Carboxy-5-fluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 3-Carboxy-5-fluorobenzeneboronic acid pinacol ester

17322-25G 17322-5G
76908-890EA 559.1 USD
76908-890 76908-892
3-Carboxy-5-fluorophenylboronic acid pinacol ester ≥97%
3-Carboxy-5-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BFO₄
Storage Temperature: Ambient
MDL Number: MFCD12756465
CAS Number: 936728-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR