3-Benzyloxy-5-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Benzyloxy-5-fluorobenzeneboronic acid

17299-25G 17299-5G
76908-866EA 418 USD
76908-866 76908-868
3-Benzyloxy-5-fluorophenylboronic acid ≥97%
3-Benzyloxy-5-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BFO₃
Storage Temperature: Ambient
MDL Number: MFCD07363782
CAS Number: 850589-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR