3-Iodo-N-methylpyridin-2-amine ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Iodo-2-(methylamino)pyridine

17269-1G 17269-25G 17269-500MG 17269-5G
76908-840EA 225.98 USD
76908-840 76908-842 76908-844 76908-846
3-Iodo-N-methylpyridin-2-amine ≥97% research grade
3-Iodo-N-methylpyridin-2-amine
Formula: C₆H₇IN₂
MW: 234.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22056741
CAS Number: 113975-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR