4-Bromo-2,6-dimethylbenzenecarbonitrile ≥97%

Supplier: AOBChem USA

Warning

17245-1G 17245-25G 17245-500MG 17245-5G
76908-830EA 135.8 USD
76908-830 76908-832 76908-834 76908-836
4-Bromo-2,6-dimethylbenzenecarbonitrile ≥97%
4-Bromo-2,6-dimethylbenzenecarbonitrile
Formula: C₉H₈BrN
Storage Temperature: Refrigerator
MDL Number: MFCD01463960
CAS Number: 5757-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR