4-Benzyloxy-1-bromo-2-fluorobenzene ≥97% research grade

Supplier: AOBChem USA

Warning

17220-25G 17220-5G
76908-820EA 204.76 USD
76908-820 76908-822
4-Benzyloxy-1-bromo-2-fluorobenzene ≥97% research grade
4-Benzyloxy-1-bromo-2-fluorobenzene
Formula: C₁₃H₁₀BrFO
Storage Temperature: Ambient
MDL Number: MFCD09801041
CAS Number: 185346-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR