4-Bromo-2-chlorophenylboronic acid ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
Synonyms:
(4-Bromo-2-chlorophenyl)boronic acid
17146-1G
17146-25G
17146-5G
76908-786EA
99.73
USD
76908-786
76908-788
76908-790
4-Bromo-2-chlorophenylboronic acid ≥97% research grade
4-Bromo-2-chlorophenylboronic acid
Formula:
C₆H₅BBrClO₂ Storage Temperature: Ambient |
MDL Number:
MFCD13195646 CAS Number: 1046861-20-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...