3-Carboxy-5-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Borono-5-methoxybenzoic acid

17136-1G 17136-25G 17136-500MG 17136-5G
76908-770EA 293.87 USD
76908-770 76908-772 76908-774 76908-776
3-Carboxy-5-methoxyphenylboronic acid ≥97%
3-Carboxy-5-methoxyphenylboronic acid
Formula: C₈H₉BO₅
MW: 195.965 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546477
CAS Number: 1050424-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR