3-Fluoro-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

17110-25G 17110-5G
76908-760EA 289.63 USD
76908-760 76908-762
3-Fluoro-2-methoxybenzoic acid ≥97%
3-Fluoro-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₃
Storage Temperature: Ambient
MDL Number: MFCD00973972
CAS Number: 106428-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR