3,4-Difluoro-2-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

17089-1G 17089-25G 17089-500MG 17089-5G
76908-746EA 257.8 USD
76908-746 76908-748 76908-750 76908-752
3,4-Difluoro-2-methoxyphenol ≥97%
3,4-Difluoro-2-methoxyphenol
Formula: C₇H₆F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD17015349
CAS Number: 158626-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR