3-Bromo-5-fluoro-2-iodobenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

17020-1G 17020-25G 17020-5G
76908-732EA 84.88 USD
76908-732 76908-734 76908-736
3-Bromo-5-fluoro-2-iodobenzotrifluoride ≥97%
3-Bromo-5-fluoro-2-iodobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₂BrF₄I
Storage Temperature: Refrigerator
MDL Number: MFCD09800680
CAS Number: 1027511-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR