3-Chloro-2-methoxy-4-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-2-methoxypyridine-4-boronic acid, (3-Chloro-2-methoxypyridin-4-yl)boronic acid

16922-1G 16922-25G 16922-5G
76908-682EA 75.33 USD
76908-682 76908-684 76908-686
3-Chloro-2-methoxy-4-pyridineboronic acid ≥97%
3-Chloro-2-methoxy-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BClNO₃
MW: 187.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06798255
CAS Number: 957060-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR