3-Bromo-2-ethyl-6-methoxypyridine ≥95%

Supplier: AOBChem USA

Warning

16848-1G 16848-25G 16848-500MG 16848-5G
76908-656EA 260.99 USD
76908-656 76908-658 76908-660 76908-662
3-Bromo-2-ethyl-6-methoxypyridine ≥95%
3-Bromo-2-ethyl-6-methoxypyridine
Formula: C₈H₁₀BrNO
MW: 216.075 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13189500
CAS Number: 848360-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR