3-Chloro-6-fluoro-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16798-1G 16798-25G 16798-5G
76908-632EA 64.72 USD
76908-632 76908-634 76908-636
3-Chloro-6-fluoro-2-methoxybenzaldehyde ≥97%
3-Chloro-6-fluoro-2-methoxybenzaldehyde
Formula: C₈H₆ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD28522703
CAS Number: 1646821-91-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR