3-Fluoro-o-anisaldehyde ≥97%

Supplier: AOBChem USA

Warning

16733-25G 16733-5G
76908-604EA 275.84 USD
76908-604 76908-606
3-Fluoro-o-anisaldehyde ≥97%
3-Fluoro-o-anisaldehyde

Precaución: for Research Use Only

Formula: C₈H₇FO₂
Storage Temperature: Refrigerator
MDL Number: MFCD04112486
CAS Number: 74266-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR