3-Bromo-5-fluoro-2-methoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

16648-1G 16648-25G 16648-5G
76908-572EA 62.6 USD
76908-572 76908-574 76908-576
3-Bromo-5-fluoro-2-methoxybenzaldehyde ≥95%
3-Bromo-5-fluoro-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11520672
CAS Number: 1009093-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR