2-(Benzyloxy)-4-bromo-1-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-1-fluoro-2-(phenylmethoxy)benzene

16544-25G 16544-5G
76908-560EA 339.49 USD
76908-560 76908-562
2-(Benzyloxy)-4-bromo-1-fluorobenzene ≥97%
2-(Benzyloxy)-4-bromo-1-fluorobenzene
Formula: C₁₃H₁₀BrFO
MW: 281.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520671
CAS Number: 1036724-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR