3-Fluoro-4-(hydroxymethyl)benzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: [3-fluoro-4-(hydroxymethyl)phenyl]boronic acid, (3-Fluoro-4-(hydroxymethyl)phenyl)boronic acid, 3-Fluoro-4-hydroxymethylbenzeneboronic acid

16535-1G 16535-25G 16535-500MG 16535-5G
76908-548EA 135.8 USD
76908-548 76908-550 76908-552 76908-554
3-Fluoro-4-(hydroxymethyl)benzeneboronic acid ≥97%
3-Fluoro-4-(hydroxymethyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₃
MW: 169.946 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13181609
CAS Number: 1082066-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR