3-Bromo-2-methoxy-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16450-1G 16450-25G 16450-5G
76908-522EA 79.57 USD
76908-522 76908-524 76908-526
3-Bromo-2-methoxy-5-methylbenzaldehyde ≥97%
3-Bromo-2-methoxy-5-methylbenzaldehyde
Formula: C₉H₉BrO₂
MW: 229.071 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13181621
CAS Number: 1224604-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR