5-Borono-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Carboxy-4-methylphenylboronic acid

16440-1G 16440-25G 16440-500MG 16440-5G
76908-508EA 173.99 USD
76908-508 76908-510 76908-512 76908-514
5-Borono-2-methylbenzoic acid ≥97%
5-Borono-2-methylbenzoic acid
Formula: C₈H₉BO₄
MW: 179.97 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13181619
CAS Number: 1256346-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR