3-Chloro-4-hydroxy-5-methylbenzaldehy ≥97% research grade

Supplier: AOBChem USA

Warning

16375-1G 16375-25G 16375-500MG 16375-5G
76908-466EA 248.26 USD
76908-466 76908-468 76908-470 76908-472
3-Chloro-4-hydroxy-5-methylbenzaldehy ≥97% research grade
3-Chloro-4-hydroxy-5-methylbenzaldehy
Formula: C₈H₇ClO₂
MW: 170.593 g/mol
Storage Temperature: Ambient
CAS Number: 107356-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR