2,4,6-Trifluoro-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Methoxy-2,4,6-trifluorobenzaldehyde

16347-1G 16347-25G 16347-5G
76908-454EA 96.55 USD
76908-454 76908-456 76908-458
2,4,6-Trifluoro-3-methoxybenzaldehyde ≥97%
2,4,6-Trifluoro-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₅F₃O₂
MW: 190.119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06660224
CAS Number: 886499-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR