3,4-Difluoro-2-methoxyaniline ≥97%

Supplier: AOBChem USA

Warning

16208-1G 16208-25G 16208-500MG 16208-5G
76908-426EA 159.14 USD
76908-426 76908-428 76908-430 76908-432
3,4-Difluoro-2-methoxyaniline ≥97%
3,4-Difluoro-2-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₇F₂NO
MW: 159.133 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368889
CAS Number: 114076-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR