3-Bromo-2-fluoro-6-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16194-1G 16194-25G 16194-5G
76908-416EA 76.39 USD
76908-416 76908-418 76908-420
3-Bromo-2-fluoro-6-(trifluoromethyl)benzaldehyde ≥97%
3-Bromo-2-fluoro-6-(trifluoromethyl)benzaldehyde
Formula: C₈H₃BrF₄O
Storage Temperature: Refrigerator
MDL Number: MFCD11520664
CAS Number: 909186-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR