4-Borono-2,6-difluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3,5-Difluoro-4-carboxyphenylboronic acid

16043-1G 16043-25G 16043-5G
76908-384EA 64.72 USD
76908-384 76908-386 76908-388
4-Borono-2,6-difluorobenzoic acid ≥97%
4-Borono-2,6-difluorobenzoic acid
Formula: C₇H₅BF₂O₄
MW: 201.92 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13181636
CAS Number: 1029716-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR