3-Bromo-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

15964-25G 15964-5G
76908-370EA 428.61 USD
76908-370 76908-372
3-Bromo-4-iodopyridine ≥97%
3-Bromo-4-iodopyridine
Formula: C₅H₃BrIN
Storage Temperature: Refrigerator
MDL Number: MFCD11110263
CAS Number: 89167-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR