3,5-Dichloro-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

15932-25G 15932-5G
76908-360EA 286.45 USD
76908-360 76908-362
3,5-Dichloro-4-iodopyridine ≥97%
3,5-Dichloro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂Cl₂IN
MW: 273.8866 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094687
CAS Number: 343781-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR