3-Bromo-2-(phenylmethoxy)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15904-1G 15904-25G 15904-5G
76908-352EA 112.46 USD
76908-352 76908-354 76908-356
3-Bromo-2-(phenylmethoxy)phenylboronic acid ≥97%
3-Bromo-2-(phenylmethoxy)phenylboronic acid
Formula: C₁₃H₁₂BBrO₃
MW: 306.948 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28976844
CAS Number: 871125-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR