3,4-Difluoro-5-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15786-1G 15786-25G 15786-500MG 15786-5G
76908-304EA 138.98 USD
76908-304 76908-306 76908-308 76908-310
3,4-Difluoro-5-methoxyphenylboronic acid ≥97%
3,4-Difluoro-5-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD09907960
CAS Number: 925910-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR