(3-(Hydroxymethyl)-4-methylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Hydroxymethyl-4-methylphenylboronic acid

15741-1G 15741-25G 15741-5G
76908-282EA 62.6 USD
76908-282 76908-284 76908-286
(3-(Hydroxymethyl)-4-methylphenyl)boronic acid ≥97%
(3-(Hydroxymethyl)-4-methylphenyl)boronic acid
Formula: C₈H₁₁BO₃
MW: 165.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617258
CAS Number: 1451391-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR