3-Bromo-5-fluoro-4-hydroxybenzaldehyde ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
15734-25G
15734-5G
76908-264EA
283.27
USD
76908-264
76908-266
3-Bromo-5-fluoro-4-hydroxybenzaldehyde ≥97% research grade
3-Bromo-5-fluoro-4-hydroxybenzaldehyde
Formula:
C₇H₄BrFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD21606586 CAS Number: 185345-46-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...