3,5-Bibromo-4-isopropoxybenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

15727-25G 15727-5G
76908-254EA 488.02 USD
76908-254 76908-256
3,5-Bibromo-4-isopropoxybenzaldehyde ≥97% research grade
3,5-Bibromo-4-isopropoxybenzaldehyde
Formula: C₁₀H₁₀Br₂O₂
MW: 321.993 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03271346
CAS Number: 486996-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR