3-Bromo-5-chloro-4-ethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15723-1G 15723-25G 15723-5G
76908-244EA 79.57 USD
76908-244 76908-246 76908-248
3-Bromo-5-chloro-4-ethoxybenzaldehyde ≥97%
3-Bromo-5-chloro-4-ethoxybenzaldehyde
Formula: C₉H₈BrClO₂
MW: 263.516 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05863698
CAS Number: 884497-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR