3,6-Dichloro-2-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

15639-25G 15639-5G
76908-200EA 346.92 USD
76908-200 76908-202
3,6-Dichloro-2-fluorobenzoic acid ≥97%
3,6-Dichloro-2-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃Cl₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD09025382
CAS Number: 916420-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR