3-Benzyloxy-6-bromo-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15610-1G 15610-25G 15610-5G
76908-176EA 109.28 USD
76908-176 76908-178 76908-180
3-Benzyloxy-6-bromo-2-fluorobenzaldehyde ≥97%
3-Benzyloxy-6-bromo-2-fluorobenzaldehyde
Formula: C₁₄H₁₀BrFO₂
MW: 309.13 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11110257
CAS Number: 1114809-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR