3-Chloro-5-fluoro-2-methoxybenzaldehyde ≥95% research grade

Supplier: AOBChem USA

Warning

15580-1G 15580-25G 15580-500MG 15580-5G
76908-140EA 194.15 USD
76908-140 76908-142 76908-144 76908-146
3-Chloro-5-fluoro-2-methoxybenzaldehyde ≥95% research grade
3-Chloro-5-fluoro-2-methoxybenzaldehyde
Formula: C₈H₆ClFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD06246207
CAS Number: 82129-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR